missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA14679
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is specific for the ∽60 kDa Tyrosine Hydroxylase in Western blots of rat brain extracts.
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Many, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Tyrosine Hydroxylase | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04177, P07101, P24529 | |
TH | |
SDS-denatured, native rat tyrosine hydroxylase purified from pheochromocytoma | |
100 μL | |
Primary | |
Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
TH | |
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
Sheep | |
Affinity chromatography | |
RUO | |
21823, 25085, 7054 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |