Learn More
CMV UL147/vCXC2 Mouse anti-Viral, Clone: 146326, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB606
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411206
  • 420.42 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 22-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
CMV UL147/vCXC2 Monoclonal antibody MAB606 Specifically detects CMV UL147/vCXC2 in Viral samples. It is validated for Western Blotting
Spezifikation
Spezifikation
UL147/vCXC2
146326
Western Blot 1 ug/mL
Purified
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects viral CMV UL147/vCXC2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant viral (rv) CMV UL146, rvMCV type 2, rvMIP-I, rvMIP-II, or rvMIP-III is observed.
Virus
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
UL147
E. coli-derived recombinant viral CMV UL147/vCXC2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3077419
Reconstitute at 0.5 mg/mL in sterile PBS.