Learn More
V5 Tag Monoclonal Antibody (E10/V4RR), Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA5152531MG
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15253 recognizes V5 (GKPIPNPLLGLDST) tagged fusion proteins. MA5-15253 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.
V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
V5 Tag | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
E10/V4RR | |
Unconjugated | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH | |
1 mg | |
Primary | |
Antibody | |
IgG1 |