missing translation for 'onlineSavingsMsg'
Learn More

VEGF-B 186 Rat anti-Mouse, Clone: 124112, R&D Systems™

Rat Monoclonal Antibody

Marke:  RD Systems (Bio-Techne) MAB767

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411846

  • 451.12 € / 500 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

VEGF-B 186 Monoclonal antibody MAB767 Specifically detects VEGF-B 186 in Mouse samples. It is validated for Western Blotting
Spezifikation

Spezifikation

VEGF-B
124112
Western Blot 1 ug/mL
P49766.2
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 165 rhVEGF-B 186 recombinant mouse (rm) VEGF-B 167 rhVEGF-C, rmVEGF-D, rhCTGF, rhPlGF, or rhLDGF is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Vegfa
E. coli-derived recombinant mouse VEGF-B 186 Pro22-Ala207 Accession # P49766.2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7423
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only