missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517609
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
YAP1 Polyclonal antibody specifically detects YAP1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
YAP1 | |
Polyclonal | |
Unconjugated | |
YAP1 | |
65 kDa Yes-associated protein; AI325207; cb194; CH211-181P1.5-001; COB1; Protein yorkie homolog; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; transcriptional coactivator YAP1; wu:fc18c04; yap; Yap1; YAP2; Yap65; Yes associated protein 1; Yes1 associated transcriptional regulator; yes-associated protein 1; yes-associated protein 2; yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; Yki; Yorkie; yorkie homolog; zgc:158380; zYAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10413, 22601, 363014 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
31 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P46937, P46938, Q2EJA0 | |
YAP1 | |
Synthetic peptide corresponding to the human sequence of YAP | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |