missing translation for 'onlineSavingsMsg'
Learn More

YBX1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA513307

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 10587220

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with bovine, mouse, rat and Xenopus based on sequence homology.

YBX1 Polyclonal antibody specifically detects YBX1 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

YBX1
Polyclonal
Unconjugated
YBX1
1700102N10Rik; BP8; BP-8; Byb1; C79409; Cbfa; CBF-A; CCAAT-binding; CCAAT-binding transcription factor 1 subunit A; CCAAT-binding transcription factor I subunit A; CSDA2; CSDB; dbpB; DNA binding protein B; DNA-binding protein B; EF1A; EFI-A; enhancer factor 1 alpha; enhancer factor 1 subunit A; enhancer factor I subunit A; major histocompatibility complex, class II, Y box-binding protein I; MDR NF1; MDR-NF1; MGC104858; MGC110976; MGC117250; Msy1; Msy-1; mYB-1a; Nsep1; NSEP-1; nuclease sensitive element binding protein 1; nuclease-sensitive element-binding protein 1; Y box binding protein 1; Y box protein 1; Y box transcription factor; YB1; YB-1; Y-box binding protein 1; Y-box transcription factor; Y-box-binding protein 1; YBX1
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
4904
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Western Blot
1.8 mg/mL
PBS with 0.09% sodium azide
P67809
YBX1
KLH conjugated synthetic peptide between 276-305 amino acids from the C-terminal region of human YBX1
400 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote