missing translation for 'onlineSavingsMsg'
Learn More

YBX2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA530614

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13495107

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: IMR32, mouse testis. Predicted reactivity: Mouse (90%), Rat (90%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

YBX2 Polyclonal antibody specifically detects YBX2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

YBX2
Polyclonal
Unconjugated
YBX2
Contrin; CSDA3; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein; MSY2; MSY2 homolog; RGD1305068; Y box binding protein 2; Y box protein 2; Y-box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2; Ybx2
Rabbit
Antigen affinity chromatography
RUO
51087, 53422
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.43 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q9Y2T7, Q9Z2C8
YBX2
Recombinant fragment corresponding to a region within amino acids 62 and 287 of Human YBX2
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote