missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yellow Fever Monoclonal Antibody (3576), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516501
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Specificity: Mouse anti Yellow Fever Virus antibody, clone 3576 recognises the Yellow Fever Virus. It is negative for other Flaviviruses tested as shown by ELISA.
Yellow Fever Monoclonal antibody specifically detects Yellow Fever in Virus samples. It is validated for ELISA.Spezifikation
Yellow Fever | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA | |
3576 | |
Unconjugated | |
YFV | |
whole yellow fever virus preparation | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |