missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yersinia pestis V Antigen Monoclonal Antibody (Va48), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA123088
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-23088 detects Yersinia pestis v antigen in Bacteria samples.
Yersinia pestis V Antigen Monoclonal antibody specifically detects Yersinia pestis V Antigen in Bacteria samples. It is validated for ELISA, Western Blot.Spezifikation
Yersinia pestis V Antigen | |
Monoclonal | |
4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
Va48 | |
Unconjugated | |
Bubonic Plague; Pasturella pestis; yersinia pseudotuberculosis | |
Recombinant full length protein. | |
100 μg | |
Primary | |
4° C | |
Liquid |