missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YES1 Monoclonal Antibody (3C6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA520232
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: MGCIKSKENK SPAIKYRPEN TPEPVSTSVS HYGAEPTTVS PCPSSSAKGT AVNFSSLSMT PFGGSSGVTP FGGASSSFSV
YES1 Monoclonal antibody specifically detects YES1 in Human samples. It is validated for ELISA, Proximity Ligation Assay (PLA), Western Blot.Spezifikation
YES1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P07947 | |
Yes1 | |
YES1 (AAH48960, 1 a.a. ∽ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Proximity Ligation Assay, Western Blot | |
3C6 | |
Unconjugated | |
Yes1 | |
cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase | |
Mouse | |
Protein A | |
RUO | |
7525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |