Learn More
YES1 Monoclonal Antibody (2F3E6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515386
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15386 targets YES1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15386 immunogen is purified recombinant fragment of YES (aa10-193) expressed in E. Coli.
YES1 Monoclonal antibody specifically detects YES1 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
YES1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P07947 | |
Yes1 | |
Purified recombinant fragment of YES (aa10-193) expressed in E. Coli. | |
RUO | |
7525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
2F3E6 | |
Unconjugated | |
Yes1 | |
cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |