missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YME1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524808
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology.
YME1L1 Polyclonal antibody specifically detects YME1L1 in Hamster, Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
YME1L1 | |
Polyclonal | |
Unconjugated | |
YME1L1 | |
ATP-dependent metalloprotease FtsH1; ATP-dependent metalloprotease FtsH1 homolog; ATP-dependent metalloprotease YME1L1; ATP-dependent metalloprotease YME1L1-like protein; ATP-dependent zinc metalloprotease YME1L1; ATP-dependent zinc metalloprotease YME1l1-like protein; FTSH; FTSH1; MEG4; meg-4; PAMP; Presenilin-associated metalloprotease; UNQ1868/PRO4304; YME1 like 1 ATPase; YME1L; YME1L1; YME1-like 1 (S. cerevisiae); YME1-like 1 ATPase; YME1-like protein 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100770453, 10730 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q96TA2 | |
YME1L1 | |
KLH conjugated synthetic peptide between 191-219 amino acids from the N-terminal region of human YMEL1 | |
400 μL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG |