Learn More
MAP3K19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529285
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAP3K19 Polyclonal antibody specifically detects MAP3K19 in Human samples. It is validated for Western Blot.Spezifikation
MAP3K19 | |
Polyclonal | |
Unconjugated | |
MAP3K19 | |
MAP3K19; Mitogen-activated protein kinase kinase kinase 19; RCK; regulated in COPD kinase; regulated in COPD, protein kinase; SPS1/STE20-related protein kinase YSK4; yeast Sps1/Ste20-related kinase 4; YSK4; YSK4 Sps1/Ste20-related kinase homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q56UN5 | |
MAP3K19 | |
Recombinant fragment corresponding to a region within amino acids 1013 and 1227 of Human YSK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |