missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545722
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GGLPGIDLSD PKQLAEFTKV KPKRSKGEPP KTVPCSYSGC EKMFRDYAAM Sequence homology: Human: 100%
YY2 Polyclonal antibody specifically detects YY2 in Human samples. It is validated for Western Blot.Spezifikation
YY2 | |
Polyclonal | |
Unconjugated | |
YY2 | |
transcription factor yin yang 2; transcription factor YY2; Yin and yang 2; YY2; YY-2; YY2 transcription factor; zinc finger protein 631; ZNF631 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
404281 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O15391 | |
YY2 | |
Synthetic peptide directed towards the C-terminal of human YY2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |