missing translation for 'onlineSavingsMsg'
Learn More

ZAP-70 Kinase Mouse, Unlabeled, Clone: 29, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610240

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15805359

  • 434.00 € / 1 Set

Verfügbar ab
16-10-2020

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

ZAP-70 is a protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP-70 contains two SH2-like domains with the PTK domain being at the C-terminus. It appears that both ZAP-70 and Syk are recruited to the phosphorylated CD3 and ζ subunits after TCR stimulation. The src-family PTKs, lck, and fyn, seem to play a role upstream of ZAP-70 and Syk where they phosphorylate CD3 and ζ. Thus both Src- and ZAP-70/Syk-family PTKs are critical for efficient induction of tyrosine phosphoproteins in heterologous cells. The significance of ZAP-70 in mediating TCR signal transduction has been confirmed by showing that ZAP-70 activity is absent in an autosomal recessive form of severe combined immunodeficiency (SCID). This is due to mutations affecting the ZAP-70 kinase domain which affect the stability of the protein. The levels of lck and fyn appear to be normal in the same cells. Results of the ZAP-70 mutations in these patients include an unusual absence of peripheral CD8+ T cells and an abundance of CD4+ T cells in peripheral lymphoid organs. These results indicate that these two populations of T cells develop through utilization of different intracellular signaling pathways.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Spezifikation

Spezifikation

ZAP-70 Kinase
29
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Videos
Dokumentation

For Research Use Only.