Learn More
APOD Mouse anti-Human, CoraLite 594, Clone: 1C6D10, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66215-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Apolipoprotein D (ApoD) is a member of the lipocalin superfamily of ligand transporters, and has been implicated in the transport of small hydrophobic molecules. ApoD is also a component of plasma high-density lipoproteins (HDL). Alteration of ApoD expression has been linked to multiple neurological disorders, including Alzheimer's disease.
Spezifikation
APOD | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG1 |
Immunofluorescence | |
1C6D10 | |
CoraLite 594 | |
100 μL | |
347 | |
Liquid |