missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APP/Protease Nexin II Pan Specific Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | APP |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Anwendungen | Western Blot, Immunohistochemistry |
Klassifikation | Polyclonal |
Beschreibung
APP Polyclonal specifically detects APP in Human samples. It is validated for Western Blot, Immunohistochemistry.Spezifikation
APP | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II | |
APP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_958817 | |
351 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP 695 +1 frameshift mutant Leu18-Arg336 Accession # NP_958817 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human APP695 +1 and human APP770 in direct ELISAs and Western blots. This antibody recognizes a common epitope present on APP695 +1 and APP770 found within the segments Leu18 - Arg288 or Pro365 - Arg411 of APP770 (Accession P05067). |
For Research Use Only