Learn More
Arginase 1 Monoclonal Antibody (A1exF5), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 12-3697-82
Weitere Details : Netto-Gewicht : 0.00100kg
Beschreibung
Description: The monoclonal antibody A1exF5 recognizes both human and mouse Arginase 1, a cytosolic enzyme (Arg1). This A1exF5 clone is compatible with both the standard intracellular protocols and the Foxp3/Transcription Factor Staining Buffer Set. Applications Reported: This A1exF5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1exF5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Arginase-1 (Arg1) is a 35 kDa enzyme converting L-arginine to urea and L-ornithine, which is the final step in the urea cycle. The resulting polyamines are important for cell proliferation and removal of toxins that arise from protein degradation. By degrading arginine, Arginase 1 deprives NO synthase of its substrate and down-regulates nitric oxide production. In both human and mouse, Arginase 1 is expressed in the liver, neutrophils, myeloid derived suppressor cells (MDSC) and neural stem cells. In human, expression in blood neutrophils but not in CCR3+ granulocytes has been reported. In mice, expression of Arginase 1 is one of the hallmarks of alternatively activated macrophages (M2a). Arginase-1 may be expressed in the myeloid cells infiltrating tumors, and is typically found in the majority of hepatocellular carcinomas. Defects in Arginase 1 are the cause of argininemia, an autosomal recessive disorder characterized by hyperammonemia.Spezifikation
Arginase 1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P05089, Q61176 | |
ARG1 | |
E.coli-derived recombinant mouse Arginase 1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
A1exF5 | |
PE | |
ARG1 | |
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
Rat | |
Affinity chromatography | |
RUO | |
11846, 383 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |