missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550114
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ARHGEF11 protein.
ARHGEF11 Polyclonal antibody specifically detects ARHGEF11 in Human samples. It is validated for Western Blot.Spezifikation
ARHGEF11 | |
Polyclonal | |
Unconjugated | |
ARHGEF11 | |
Arhgef11; B930073M02; E130307F09; glutamate transporter EAAT4 associated protein 48; glutamate transporter EAAT4-associated protein 48; Gtrap48; KIAA0380; mKIAA0380; PDZ-RhoGEF; Prg; Rho guanine exchange factor (GEF) 11; Rho guanine nucleotide exchange factor (GEF) 11; rho guanine nucleotide exchange factor 11; RhoA-specific guanine nucleotide exchange factor; RhoGEF; RhoGEF glutamate transport modulator; RhoGEF glutamate transport modulator GTRAP48; RP11-356J7.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9826 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15085 | |
ARHGEF11 | |
Synthesized peptide derived from N-terminal of human ARHGEF11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |