missing translation for 'onlineSavingsMsg'
Learn More

ARL2BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10090-2-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886243

  • 399.00 € / 150 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2. GTP with high affinity but does not interact with ARL2. GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2. GTP but lack of ARL2 GTPase-activating protein activity.
Spezifikation

Spezifikation

ARL2BP
Polyclonal
Unconjugated
ARL2BP
ARF like 2 binding protein, ARL2BP, BART, BART1, Binder of ARF2 protein 1
Rabbit
Antigen Affinity Chromatography
RUO
107566, 23568, 498910
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4V8C5, Q9D385, Q9Y2Y0
Arl2bp
ARL2BP Fusion Protein Ag0138
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote