missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26223-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is the human ortholog of the Drosophila melanogaster ′abnormal spindle′ gene (asp), which is essential for normal mitotic spindle function in embryonic neuroblasts. Studies in mouse also suggest a role of this gene in mitotic spindle regulation, with a preferential role in regulating neurogenesis. Mutations in this gene are associated with microcephaly primary type 5. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
ASPM | |
Polyclonal | |
Unconjugated | |
ASPM | |
ASP, Asp homolog, ASPM, Calmbp1, MCPH5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
259266 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IZT6 | |
ASPM | |
ASPM Fusion Protein Ag24166 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |