Learn More
ATP1A1-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Alle ansehen Proteintech ProdukteBeschreibung
The ATP1A1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ATP1A1-Specific. This antibody recognizes human, mouse antigen. The ATP1A1-Specific antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Spezifikation
Spezifikation
| Antigen | ATP1A1-Specific |
| Anwendungen | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Konzentration | 1000 μg/mL |
| Konjugat | CoraLite Plus 488 |
| Gensymbole | ATP1A1, Sodium pump subunit alpha 1 |
| Wirtsspezies | Rabbit |
| Menge | 20 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 476 |
| Zielspezies | Human, Mouse |
| Mehr anzeigen |