missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16143-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation.Spezifikation
ATP6V1G1 | |
Polyclonal | |
Unconjugated | |
ATP6V1G1 | |
ATP6G, ATP6G1, ATP6GL, ATP6J, ATP6V1G1, V ATPase 13 kDa subunit 1, V ATPase subunit G 1, Vma10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
298103, 66290, 9550 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75348, Q9CR51 | |
Atp6v1g1 | |
ATP6V1G1 Fusion Protein Ag9217 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |