missing translation for 'onlineSavingsMsg'
Learn More

ATPA, Tocris Bioscience™

Selective, potent GluR5 agonist

Marke:  Tocris (Bio-Techne) 110750

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418543

  • 1011.15 € / 50 Milligramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

A selective and potent GluR5 kainate receptor agonist (Ki = 4.3 nM), inactive at GluR6 (Ki > 1 mM) and only weakly active at AMPA receptors (GluR1-4) and the kainate receptors KA-2 and GluR7 (Ki values of 6 - 14 μM). Also available as part of the Kainate Receptor Tocriset™ .
Spezifikation

Chemische Identifikatoren

140158-50-5
228.248
atpa, atpa-tert, r,s-atpa, alpha-amino-3-hydroxy-5-tert-butyl-4-isoxazolepropionate, 2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propionic acid, rs-2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo-4-isoxazolepropanoic acid, 4-isoxazolepropanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo, atpa, solid, acmc-20cirp
2-amino-3-(5-tert-butyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
C10H16N2O4
PIXJURSCCVBKRF-UHFFFAOYSA-N
2253
CC(C)(C)C1=C(C(=O)NO1)CC(C(=O)O)N

Spezifikation

ATPA
140158-50-5
atpa, atpa-tert, r,s-atpa, alpha-amino-3-hydroxy-5-tert-butyl-4-isoxazolepropionate, 2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propionic acid, rs-2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo-4-isoxazolepropanoic acid, 4-isoxazolepropanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo, atpa, solid, acmc-20cirp
CC(C)(C)C1=C(C(=O)NO1)CC(C(=O)O)N
228.248
228.25
50mg
C10H16N2O4
PIXJURSCCVBKRF-UHFFFAOYSA-N
2-amino-3-(5-tert-butyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
2253
>98%
Videos