missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
ATRX Polyclonal specifically detects ATRX in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spezifikation
Spezifikation
| Antigen | ATRX |
| Anwendungen | Immunocytochemistry, Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL |
| Zusammensetzung | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gen-Alias | alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX |
| Gensymbole | ATRX |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EFRAMDAVNKEKNTKEHKVIDAKFETKARKGEKPCALEKKDISKSEAKLSRKQVDSEHMHQNVPTEEQRTNKSTGGEHKKSDRKEEPQYEPANTSE |
| Mehr anzeigen |
For Research Use Only
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?