missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ X50 1,1ML SQ 96 DEEPWELL PLATE
For sample collection, in-vitro growth chambers and long term storage.
Marke: Axygen™ P-1ML-SQ-C
Weitere Details : Netto-Gewicht : 2.70000kg
Beschreibung
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.Spezifikation
ANSI, SLAS 1-2004 | |
Without Lid | |
Autoclavable | |
Deep-Well Square Plate | |
1.4 mL | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
5/Pk. |
Standard | |
8 x 12 | |
Clear | |
Polypropylene | |
96 | |
Square | |
50/Cs. | |
10/Cs. |