missing translation for 'onlineSavingsMsg'
Learn More

AZ 10606120 dihydrochloride, Tocris Bioscience™

Potent P2X7 receptor antagonist

Marke:  Tocris (Bio-Techne) 332310

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412784

  • 237.61 € / 10 Milligramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent P2X 7 receptor antagonist (K D values are 1.4 and 19 nM at human and rat P2X 7 receptors respectively). Binds in a positive cooperative manner to sites distinct from, but coupled to, the ATP binding site and acts as a negative allosteric modulator. Inhibits tumor growth and displays antiangiogenic effects in mice.
Spezifikation

Chemische Identifikatoren

607378-18-7
495.489
az 10606120 dihydrochloride, n-2-2-2-hydroxyethyl amino ethyl amino-5-quinolinyl-2-tricyclo 3.3.1.13,7 dec-1-ylacetamide dihydrochloride, 2-1-adamantyl-n-2-2-2-hydroxyethyl amino ethyl amino quinolin-5-yl acetamide dihydrochloride
2-(1-adamantyl)-N-[2-[2-(2-hydroxyethylamino)ethylamino]quinolin-5-yl]acetamide;dihydrochloride
C25H36Cl2N4O2
BVFONFUUWORSPO-UHFFFAOYSA-N
56972227
C1C2CC3CC1CC(C2)(C3)CC(=O)NC4=CC=CC5=C4C=CC(=N5)NCCNCCO.Cl.Cl

Spezifikation

AZ 10606120 dihydrochloride
607378-18-7
az 10606120 dihydrochloride, n-2-2-2-hydroxyethyl amino ethyl amino-5-quinolinyl-2-tricyclo 3.3.1.13,7 dec-1-ylacetamide dihydrochloride, 2-1-adamantyl-n-2-2-2-hydroxyethyl amino ethyl amino quinolin-5-yl acetamide dihydrochloride
C1C2CC3CC1CC(C2)(C3)CC(=O)NC4=CC=CC5=C4C=CC(=N5)NCCNCCO.Cl.Cl
495.489
495.48
10mg
C25H36Cl2N4O2
BVFONFUUWORSPO-UHFFFAOYSA-N
2-(1-adamantyl)-N-[2-[2-(2-hydroxyethylamino)ethylamino]quinolin-5-yl]acetamide;dihydrochloride
56972227
>98%
Videos