Learn More
Synergy Healthcare™ X12 Azowipe Hard Surface Bactericidal Wipes
Kills most pathogenic micro-organisms including E.Coli, Staphylococcus, MRSA, Hepatitis B, Hepatitis C, HIV, and many others. Synergy Healthcare™ Azowipe™ Hard Surface Disinfecting Wipes are widely used in both health care areas as well as in food manufacturing and educational settings.
Marke:  Synergy Healthcare™ 81103
Weitere Details : CAS : 67-63-0 Netto-Gewicht : 2.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Artikelnummer. 15611944
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
  • 70% Isopropyl alcohol disinfectant
  • Broad spectrum activity against a range of micro-organisms
  • Extremely fast contact times from 15 seconds
  • Non-smearing and streak-free when used on hard surfaces.
  • Certified Halal approved by European Halal Development agency.
  • Suitable for food manufacturing, hospitality and catering sectors.
  • Compliant with HTM-01-05 guidelines