Learn More
Synergy Healthcare™ X12 Azowipe Hard Surface Bactericidal Wipes
Kills most pathogenic micro-organisms including E.Coli, Staphylococcus, MRSA, Hepatitis B, Hepatitis C, HIV, and many others. Synergy Healthcare™ Azowipe™ Hard Surface Disinfecting Wipes are widely used in both health care areas as well as in food manufacturing and educational settings.
Marke: Synergy Healthcare™ 81103
Weitere Details : CAS : 67-63-0 Netto-Gewicht : 2.40000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Beschreibung
- 70% Isopropyl alcohol disinfectant
- Broad spectrum activity against a range of micro-organisms
- Extremely fast contact times from 15 seconds
- Non-smearing and streak-free when used on hard surfaces.
- Certified Halal approved by European Halal Development agency.
- Suitable for food manufacturing, hospitality and catering sectors.
- Compliant with HTM-01-05 guidelines