Learn More
B4GALNT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA553371
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LLPEERLRNL FSYDGIWLFP KNQCKCEANK EQGGYNFQDA YGQSDLPAVK ARRQAEFEHF QRREGLPRPL PLLVQPNLPF GYPVHGVEVM PLHTVPIPGL QFEGPDA Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 25%.
B4GALNT2 Polyclonal antibody specifically detects B4GALNT2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
B4GALNT2 | |
Polyclonal | |
Unconjugated | |
B4GALNT2 | |
AI593864; B4galnt2; B4GALT; beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-1,4-N-acetyl-galactosaminyltransferase 2; beta-4-N-acetylgalactosaminyltransferase; CT-Gal NAc transferase; Dlb1; Dlb-1; dolichos lectin binding 1; Galg2; Galgt2; Ggm3; Igf2bp1; N-acetylgalactosaminyltransferase; sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosyl-N-acetylglucosaminylpolypeptide-beta-1, 4-N-acetylgalactosaminyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NHY0 | |
B4GALNT2 | |
Recombinant protein corresponding to Human B4GALNT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only