missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA568113
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse and Rat samples.
B4GALT6 Polyclonal antibody specifically detects B4GALT6 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
B4GALT6 | |
Polyclonal | |
Unconjugated | |
B4GALT6 | |
AA536803; AU022389; B4galt6; B4Gal-T6; beta-1, 4-galactosyltransferase VI; beta-1,4-galactosyltransferase 6; Beta-1,4-GalTase 6; beta4Gal-T6; beta4GalT-VI; GalT; Glucosylceramide beta-1,4-galactosyltransferase; LacCer synthase; Lactosylceramide synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 6; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 6; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6; UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6; zgc:55730 | |
Rabbit | |
affinity chromatography | |
RUO | |
56386, 65196, 9331 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O88419, Q9UBX8, Q9WVK5 | |
B4GALT6 | |
Synthesized peptide derived from human B4GALT6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |