Learn More
B9D2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA559923
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: WSQDSFGRCQ LAGYGFCHVP SSPGTHQLAC PTWRPLGSWR EQLARAFVGG GPQLLHGDTI YSGADRYRLH TAAGGTVHLE IGLLLRNFDR YG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
B9D2 Polyclonal antibody specifically detects B9D2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
B9D2 | |
Polyclonal | |
Unconjugated | |
B9D2 | |
B9 domain-containing protein 2; B9 protein domain 2; B9D2; ICIS-1; involved in cIlia stability-1; MKS10; MKS1-related protein 2; MKSR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80776 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BPU9 | |
B9D2 | |
Recombinant protein corresponding to Human B9D2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only