Learn More
Bode Chemie™ Bacillol™ Tissues
Product Code. 16552852
Change view
Click to view available options
Packaging Quantity:
100 Wipes
80 Wipes
Container Type:
Pop-up canister
Unit Size:
Pack of 100
Pack of 80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Container Type Unit Size
16552852 100 Wipes Pop-up canister Pack of 100
16210991 80 Wipes Pop-up canister Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16552852 Supplier Bode Chemie™ Supplier No. 9805031
Quantity Price
1 19.01 € / 100 Stück
12 18.10 € / 100 Stück
 Restricted Item
Lager vorrätig
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Bacillol™ Tissues - Alcohol-based rapid disinfectant wipes in a practical dispenser.
  • Ready-to-use disinfectant wipes
  • Quickly and comprehensively effective
  • Broad material compatibility with alcohol-resistant surfaces
  • Aldehyde, dye and perfume free
  • Easy removal from practical dispenser box
  • Refillable
  • Required spectrum of activity SARS-CoV-2: limited virucidal activity
Specifications
Product Type Disinfectant Wipes
Container Type Pop-up canister
Disposable Single-use
For Use With (Application) Disinfection of Alcohol-Resistant Surfaces and Medical Devices
Packaging Quantity 100 Wipes