Learn More
BAG6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA562770
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SDAYLSGMPA KRRKTMQGEG PQLLLSEAVS RAAKAAGARP LTSPESLSRD LEAPEVQESY RQQLRSDIQK RLQEDPNYS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
BAG6 Polyclonal antibody specifically detects BAG6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
BAG6 | |
Polyclonal | |
Unconjugated | |
Bag6 | |
2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7917 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P46379 | |
Bag6 | |
Recombinant protein corresponding to Human BAG6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only