Learn More
BANF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA560062
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FLSEPIGEKD VCWVDGISHE LAINLVTKGI NKAYILLGQF LLMHKNEAEF QRWLICCFGA TECEAQQTSH CLKEWCA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
BANF2 gene ontology annotations related to this gene include DNA binding.Spezifikation
BANF2 | |
Polyclonal | |
Unconjugated | |
BANF2 | |
4930517K23Rik; BAF2; BAFL; BAF-L; Baf-like; BANF family member 2; Banf2; barrier to autointegration factor 2; barrier-to-autointegration factor 2; Barrier-to-autointegration factor-like protein; C20orf179; Gm115; likely ortholog of H. sapiens Chromosome 20 open reading frame 179 (C20orf179); LOC538599 protein; RGD1307909; testis secretory sperm-binding protein Li 212a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H503 | |
BANF2 | |
Recombinant protein corresponding to Human BANF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only