missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Underbench lab models:
- Delivery kerbside drop to end-user or to dealer ramp
- Mainland only (excluding islands and to valley station only)
Specifications
Specifications
| For Use With (Equipment) | Table top/ Under bench |
| Product Type | Premium delivery |
| Duration | 2 Weeks |
| Target Audience | Research Laboratories |
| Species | All |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?