missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Basic Delivery Service Upright Lab Models

Product Code. 30281202 Shop All Liebherr Appliances Products
Change view
Click to view available options
Duration:
2 Weeks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
30281202 2 Weeks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281202 Supplier Liebherr™ Supplier No. Delivery_Prem_Standgeräte
 Restricted Item
Verfügbar ab: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Basic Delivery Service Upright Lab Models, curbside drop to end-user or to dealer ramp.

  • Upright lab models with polystyrol interior:
  • Delivery kerbside drop to end-user or to dealer ramp
  • Mainland only (excluding islands and to valley station only)

Specifications

For Use With (Equipment) Uprights
Duration 2 Weeks
Product Type Premium delivery
Target Audience Research Laboratories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.