Learn More
BAZ1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA551736
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RGHRESALKE TLLQEKSRIC AQLARFSEEK FHFSDKPQPD SKPTYSRGRS SNAYDPSQMC AEKQLELRLR DFLLDIEDRI YQGTLGAIKV TDRHIWRSAL ESGRYELLSE ENKENGIIKT VNEDVEEMEI DEQTK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
BAZ1A Polyclonal antibody specifically detects BAZ1A in Human samples. It is validated for Immunohistochemistry.Spezifikation
BAZ1A | |
Polyclonal | |
Unconjugated | |
BAZ1A | |
ACF1; ATP-dependent chromatin remodeling protein; ATP-dependent chromatin-remodeling protein; ATP-utilizing chromatin assembly and remodeling factor 1; B930060C03; Baz1a; BC065123; bromodomain adjacent to zinc finger domain 1A; bromodomain adjacent to zinc finger domain protein 1A; bromodomain adjacent to zinc finger domain, 1A; cbp146; CHRAC subunit ACF1; gene trap locus 5; Gtl5; hACF1; HSPC317; hWALp1; WALp1; WCRF180; Williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11177 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRL2 | |
BAZ1A | |
Recombinant protein corresponding to Human BAZ1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only