Learn More
BBX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552246
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NSLPQYSPVT FDRKCVPVPR KKKKTGNVSS EPTKTSKGSG DKWSNKQLFL DAIHPTEAIF SEDRNTMEPV HKVKNIPSIF NTPEPTTTQE PLVGSQKRKA RKTKI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
BBX Polyclonal antibody specifically detects BBX in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
BBX | |
Polyclonal | |
Unconjugated | |
BBX | |
5530401J07Rik; 5730403O13Rik; Ag recognized by Treg cells 1; ARTC1; BBX; BBX high mobility group box domain containing; BBX, HMG-box containing; bobby sox homolog; bobby sox homolog (Drosophila); HBP2; HMG box transcription factor BBX; HMG box-containing protein 2; HMG-BOX transcription factor BBX; HSPC339; MDS001; x 001 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WY36 | |
BBX | |
Recombinant protein corresponding to Human BBX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only