Learn More
BCLAF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552320
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LLDRGNTRDK EASKEKGSEK GRAEGEWEDQ EALDYFSDKE SGKQKFNDSE GDDTEETEDY RQFRKSVLAD QGKSFATASH RNTEEEGLKY KSKVSLKGNR ESDGFREEKN YKLKETGYVV ERPSTTKDKH KEED Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
BCLAF1 Polyclonal antibody specifically detects BCLAF1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
BCLAF1 | |
Polyclonal | |
Unconjugated | |
BCLAF1 | |
2610102K23Rik; 2700025J07Rik; 2810454G14Rik; 5730534O06Rik; Aa2-041; AI450190; AW556225; BCL2 associated transcription factor 1; bcl-2-associated transcription factor 1; BCL2-associated transcription factor 1; Bcl-2-associated transcription factor 1-like protein; BCLAF1; BCLAF1 and THRAP3 family member 1; bK211L9.1; BP-1; BTF; Btf-s; KIAA0164; mKIAA0164 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYF8 | |
BCLAF1 | |
Recombinant protein corresponding to Human BCLAF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only