Learn More
BD H2Db:Ig Recombinant Fusion Protein-Dimeric, Mouse
For immunoassay protocols
Marke: BD 551323
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The H-2D[b]:Ig fusion protein consists of three extracellular major histocompatibility complex (MHC) class I H-2D[b] domains that are fused to VH regions of mouse IgG1.
- Format: Purified
- Clone: DimerX/H-2Db
- Isotype: Mouse IgG1, λ
- Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide.
Flow Cytometry, Enzyme Linked Immunosorbent Assay
Spezifikation
0.5mg/mL | |
Aqueous Buffered solution containing 0.09% Sodium Azide | |
7.2 | |
0.25mg | |
Recombinant |
ELISA, Flow Cytometry | |
Mouse H-2Db:Ig Recombinant Fusion Protein | |
Affinity Chromatography, SDS-PAGE | |
4°C | |
Solution |
Research Use Only