missing translation for 'onlineSavingsMsg'
Learn More

BD Intramedic™ Luer-Stub Adapters

Product Code. 17967282
Change view
Click to view available options
Size:
18 G
20 G
Color:
Pink
Yellow
For Use With (Equipment):
Intramedic™ PE100 and 90
Intramedic™ PE190 and 160
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color For Use With (Equipment) Unit Size
17967282 20 G Yellow Intramedic™ PE100 and 90 Pack of 100
15915570 18 G Pink Intramedic™ PE190 and 160 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17967282 Supplier BD Supplier No. 427564

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fits Intramedic tubings

  • Sterile

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
For Use With (Equipment) Intramedic™ PE100 and 90
Size 20 G
Product Type Adapter
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.