missing translation for 'onlineSavingsMsg'
Learn More

Belotecan-d7-Hydrochlorid, TRC

Artikelnummer. 30345084
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30345084 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30345084 Supplier Toronto Research Chemicals Supplier No. TRCB13130210MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Belotecan-d7 Hydrochloride
CAS 1346598-22-8
Summenformel C25H21D7ClN3O4
Menge 10 mg
Synonym (4S)-4-Ethyl-4-hydroxy-11-[2-[(1-methylethyl-d7)amino]ethyl]-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione Hydrochloride, CKD 602-d7
InChI-Formel InChI=1S/C25H27N3O4.ClH/c1-4-25(31)19-11-21-22-17(12-28(21)23(29)18(19)13-32-24(25)30)15(9-10-26-14(2)3)16-7-5-6-8-20(16)27-22;/h5-8,11,14,26,31H,4,9-10,12-13H2,1-3H3;1H/t25-;/m0./s1/i2D3,3D3,14D;
SMILES Cl.[2H]C([2H])([2H])C([2H])(NCCc1c2CN3C(=O)C4=C(C=C3c2nc5ccccc15)[C@@](O)(CC)C(=O)OC4)C([2H])([2H])[2H]
IUPAC-Name (19S)-19-Ethyl-10-[2-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)ethyl]-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2,4,6,8,10,15(20)-heptaen-14,18-dion;hydrochlorid
Molekulargewicht (g/mol) 477
Formelmasse 476.2208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.