missing translation for 'onlineSavingsMsg'
Learn More

Beta-Nicotinamid-adenin-dinucleotid, Affymetrix/USB™

Catalog Number 15805138
Click to view available options
Menge:
1 g
5 g
10 g
25 g
100 g
Verpackung:
Kunststoffflasche
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53-84-9
C21H27N7O14P2
663.43
MFCD00150377
BAWFJGJZGIEFAR-WIWLTUSXNA-N
nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
15938971
CHEBI:57540
[[(2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methylphosphat
NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53-84-9
C21H27N7O14P2
663.43
MFCD00150377
BAWFJGJZGIEFAR-WIWLTUSXNA-N
nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
15938971
CHEBI:57540
[[(2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methylphosphat
NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material beta-Nicotinamide Adenine Dinucleotide
Verpackung Kunststoffflasche
Menge 10 g
Beilstein 3584133
Formelmasse 663.43 (Anhydrous)
Physikalische Form Pulver
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'