missing translation for 'onlineSavingsMsg'
Learn More

Betaine, 5M Solution, Molecular Biology Grade, Ultrapure

Product Code. 17729629
Change view
Click to view available options
Quantity:
10 mL
5 x 1.5 mL
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
10mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
17729629 5 x 1.5 mL Plastic bottle -
15855428 10 mL Plastic bottle 10mL
15865428 100 mL Plastic bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17729629 Supplier Thermo Scientific Alfa Aesar Supplier No. J77507.V^
 Restricted Item
Lager vorrätig
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-43-7 | C5H11NO2 | 117.148 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00012123
InChI Key KWIUHFFTVRNATP-UHFFFAOYSA-N
Synonym betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin
PubChem CID 247
ChEBI CHEBI:17750
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

Packaging Plastic bottle
Quantity 5 x 1.5 mL
Beilstein 3537113
Grade Molecular Biology, Ultrapure
Physical Form Solution
Chemical Name or Material Betaine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.