Learn More
BHLHB5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA564189
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EDDLFLHKSL SASTSKRLEA AFRSTPPGMD LSLAPPPRER PASSSSSPLG CFEPADPEGA GLLLPPP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
BHLHB5 Polyclonal antibody specifically detects BHLHB5 in Human samples. It is validated for Immunohistochemistry.Spezifikation
BHLHB5 | |
Polyclonal | |
Unconjugated | |
BHLHE22 | |
basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family member e22; basic helix-loop-helix family, member e22; Beta3; BHLHB5; bHLHe22; CAGL85; Class B basic helix-loop-helix protein 5; class E basic helix-loop-helix protein 22; Protein BETA3; TNRC20; trinucleotide repeat containing 20; trinucleotide repeat-containing gene 20 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.16 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFJ8 | |
BHLHE22 | |
Recombinant protein corresponding to Human BHLHB5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only