Learn More
Invitrogen™ Bimagrumab Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA541992
Descrizione
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Bimagrumab has been used in trials studying the treatment and supportive care of Sarcopenia, Skeletal Muscle, Mechanical Ventilation, Sporadic Inclusion Body Myositis, and Sporadic Inclusion Body Myositis (sIBM), among others.
Specifica
| Bimagrumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BYM338 | |
| Human ACVR2A/ACVR2B. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, Flow Cytometry | |
| 1.99 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 λ |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.