Learn More
Tocris Bioscience™ BIMU 8
Beschreibung
BIMU 8 is a potent 5-HT4 receptor full agonist (KD = 77 nM and EC50 = 18 nM for wild type 5-HT4 receptors). Binds to receptors in a mechanism different to that of the endogenous ligand 5-HT.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 2,3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride |
| CAS | 134296-40-5 |
| Prozentgehaltsbereich | >98% |
| Menge | 5 mg |
| Summenformel | C19H26N4O2.HCl |
| Formelmasse | Observed MW: 378.9 |