Learn More
Simport™ Bioblock 96-Deep-Well-Platten aus Polypropylen
Marke: Simport™ T110-10Y
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Standard | |
Plate is used mainly for compound storage and enzyme assays | |
24/Pk. | |
96 |
Yellow | |
Polypropylene | |
2.1 mL | |
8 x 12 |