Learn More
ACROBiosystems Biotinylated Mouse BAFFR / TNFRSF13C Protein, Fc, Avitag™
Biotinylated Protein
Marke:  ACROBiosystems BAR-M82F0-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15886007
  • 1733.51 € / 200 Mikrogramm
Verfügbar ab: 06-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 35.1 kDa. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-116 kDa under no
Spezifikation
Spezifikation
NP_082351
35.1 kDa
200 µg
Ser 10 - Ala 71
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Mouse
Powder
>95% as determined by SDS-PAGE
Biotinylated Mouse BAFFR, Fc,Avitag (BAR-M82F0) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accession # NP_082351).
-20°C
BAFFR
Fc Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins