missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chlorethyl)aminophosphorsäuredichlorid, TRC

Artikelnummer. 30333154
Change view
Click to view available options
Menge:
100 mg
Unit Size:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30333154 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333154 Supplier Toronto Research Chemicals Supplier No. TRCB418810100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Bis(2-chloroethyl)aminophosphoric Dichloride
CAS 127-88-8
Summenformel C4 H8 Cl4 N O P
Menge 100 mg
Synonym Phosphoramidic dichloride, N,N-bis(2-chloroethyl)- (ACI), N,N-Bis(2-chloroethyl)phosphoramidic dichloride (ACI), Phosphoramidic dichloride, bis(2-chloroethyl)- (7CI, 8CI, 9CI), (N,N-Di-2-chloroethyl) phosphoramidic dichloride, 2-Chloro-N-(2-chloroethyl)-N-dichlorophosphorylethanamine, Bis(2-chloroethyl)aminophosphoric dichloride, Bis(2-chloroethyl)aminophosphoryl dichloride, Bis(2-chloroethyl)phosphoramidic dichloride, N,N-Bis(2-chloroethyl)phosphamide dichloride, NSC 64119
InChI-Formel InChI=1S/C4H8Cl4NOP/c5-1-3-9(4-2-6)11(7,8)10/h1-4H2
SMILES ClCCN(CCCl)P(=O)(Cl)Cl
IUPAC-Name 2-Chlor-N-(2-Chlorethyl)-N-dichlorphosphorylethanamin
Molekulargewicht (g/mol) 258.9
Formelmasse 256.9098
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.